Placas de Petri Falcon®, 50×9 mm, tapa hermética, estériles, Corning
Proveedor: Corning
351006 351008 351029 351058 351007
391-1997EA 268 EUR
391-1997 391-1998 391-2002 391-2003 391-2266
Placas de Petri Falcon®, 50×9 mm, tapa hermética, estériles, Corning
Placas de cultivo Placas de Petri
Dishes are constructed with crystal-grade polystyrene and a flat bottom surface for distortion-free optics
  • Flat, distortion free optics
  • Stacking rings allow for easier stacking and handling
  • Durable construction for stable dish manipulation
Small diameter saves media and space. Easily sealed with tape for collecting and transporting specimens. Polystyrene. Sterilized by gamma irradiation. Packed 20 per bag.