Barium niobium oxide ≥99.9% (metals basis)

Proveedor: Thermo Fisher Scientific
Atención

39130.09 39130.18
39130.09EA 76.2 EUR
39130.09 39130.18
Barium niobium oxide ≥99.9% (metals basis)
Barium niobium oxide

Purity: 99.9% CAS No.: 12009-14-2 Molecular Formula: BaNb2O6 Formula Weight: 419.11 Physical Form: -100 Mesh Powder Solubility: Insoluble in water UN No.: UN1564 MDL No.: MFCD00064751

Formula: BaNb₂O₆
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00064751
Núm. CAS: 12009-14-2
EINECS: 234-543-1
UN: 1564
ADR: 6.1,III

Order Now

ESPECIFICACIONES

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR