Protein Coated Cell Culture Dishes, CELLCOAT®, Poly-Lysine
Proveedor: Greiner Bio-One
664940_40 628930_100 628940_100
392-0046EA 370 EUR
392-0046 392-0285 392-0270
Protein Coated Cell Culture Dishes, CELLCOAT®, Poly-Lysine
Placas de cultivo Placas para cultivo celular
PS, coated with poly-lysine. Poly-D-Lysine (PDL) and Poly-L-Lysine (PLL) are synthetic molecules that are used to improve the adhesion of different cell types to polystyrene surfaces. The cultivation efficiency of individual cell lines can be improved, especially when serum-free or serum-reduced medium is used, or when experiments such as transfections are performed. As the PDL is synthetic it is free of the impurities carried by other proteins.
  • Improved cell adhesion, proliferation and growth of transfected cell lines (eg. HEK-293, PC12, L929, certain 3T3