Roller bottles, Nunc™ TufRol™
Proveedor: Thermo Fisher Scientific
181702 141744 142720 142744 182702 182720 182744 183302 183902 184302 184344 184902 184920 184944
392-0335EA 210 EUR
392-0335 734-2923 734-2924 734-2925 392-0336 392-0337 392-0338 392-0339 392-0340 392-0341 392-0342 392-0343 392-0344 392-0345
Roller bottles, Nunc™ TufRol™
Frascos Frascos Roller
PS, sterile.
  • One-piece seamless design reduces possibility of leaking through a seam
  • Shallow and deep indents allow for excellent manual and automated handling
  • Proprietary manufacturing process allows for greater impact resistance
  • Vertical expanded surfaces version facilitates emptying and reduces retention of product
Nunc™ TufRol™ have been developed for applications such as the industrial scale production of vaccines, monoclonal antibodies and biotherapeutics, where reliability and reproducibility are essential.
Precaución: Los productos de estaMore Product Information