Agua, Cell Culture Grade pyrogen-free

Proveedor: VWR Chemicals

Sinónimos: Hydrogen oxide

392-0438EA 10.12 EUR
392-0438 392-0439
Agua, Cell Culture Grade pyrogen-free
Agua

Cell culture water is high quality water suitable for use as a solvent in the preparation of cell culture media and laboratory reagents.


  • Pyrogen-free


Suitable for dissolving dry powder cell culture media in accordance with the technical data sheet.


Precaución: The product is intended to be used in vitro, in laboratory only. Do not use it in therapy, human or veterinary applications.

Formula: H₂O
MW: 18.02 g/mol
Pto. Ebullición: 100 °C (1013 hPa)
Pto. de fusión: 0 °C
Densidad: 1 g/cm³ (4 °C)
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00011332
Núm. CAS: 7732-18-5
EINECS: 231-791-2
Índice Merck: 13,10098

Order Now

Resultados test especificaciones

Color colourless, clear solution
Endotoxins < 0.005 EU/ml
Sterility test Fungi and yeasts
Shelf life 48 months


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR