Rat IgG2a Isotype Control [clone: 2A3]

Proveedor: Tonbo Biosciences
40-4321-U100
40-4321-U100EA 67.1 EUR
40-4321-U100
Rat IgG2a Isotype Control [clone: 2A3]
Anticuerpos
The 2A3 immunoglobulin is useful as an isotype-matched control. The 2A3 immunolglobulin has an unknown binding specificity and is used as an isotype control for rat IgG2a antibodies.

Type: Control
Antigen: 2A3
Clonality:
Clone: 2A3
Conjugation: In Vivo Ready™
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR