Potassium pentachlororhodate(III) (≥28,2% Rh) ≥99.99% (metals basis), Premion®

Proveedor: Thermo Fisher Scientific
Atención

40409.03 40409.06
40409.03EA 837 EUR
40409.03 40409.06
Potassium pentachlororhodate(III) (≥28,2% Rh) ≥99.99% (metals basis), Premion®
Potassium pentachlororhodate(III)

Purity: 99.99% CAS No.: 65980-75-8 Molecular Formula: K2RhCl5 Formula Weight: 358.37 Physical Form: Powder Solubility: Slightly soluble in cold water. Insoluble in alcohol MDL No.: MFCD00049661

Formula: K₂RhCl₅
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00049661
Núm. CAS: 65980-75-8
EINECS: 265-989-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR