Glicina ≥99.5% (base secada), Bioreactivo ultra puro for Electrophoresis, Liquid Chromatography and Molecular Biology, for liquid chromatography, J.T.Baker®

Proveedor: Avantor

4059-02EA 142.46 EUR
4059-02 4059-06 4059-00 4059-07
Glicina ≥99.5% (base secada), Bioreactivo ultra puro for Electrophoresis, Liquid Chromatography and Molecular Biology, for liquid chromatography, J.T.Baker®
Glicina
Formula: NH₂CH₂COOH
Pto. Ebullición: 181 °C (1013 hPa)
Pto. de fusión: 83 °C
Densidad: 1,64 g/cm³ (20 °C)
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00008131
Núm. CAS: 56-40-6
EINECS: 200-272-2
Índice Merck: 13,04504
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d944007d30d5d4650062014a1b152bd9bfb66ea3&vid=093f4227cbdd1c4c6515ef9c4b85d93799877dcc&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

For Laboratory,Research,or Manufacturing Use ≥99.5%
Assay (C₂H₅NO₂) (dried basis, by non-aqueous acid- base titration) ≥99.5 %
Homogeneity by TLC Passes Test
Residue after Ignition ≤0.10 %
Loss on Drying at 105°C ≤0.2 %
Readily Carbonizable Substances Passes Test
Absorbance of a 1M Solution - 280 nm ≤0.100
RNase Activity (None Detected) Passes Test
DNase Activity (None Detected) Passes Test
Protease Activity (None Detected) Passes Test
Arsenic (As) ≤3 ppm
ACS - Heavy Metals (as Pb) ≤5 ppm
Iron (Fe) ≤0.003 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR