Barium isopropoxide 20% (w/v) in 2-propanol

Proveedor: Thermo Fisher Scientific
Peligro

41338.AB 41338.AD
41338.ABEA 88.1 EUR
41338.AB 41338.AD
Barium isopropoxide 20% (w/v) in 2-propanol
Barium isopropoxide

CAS No.: 24363-37-9 Molecular Formula: C6H14BaO2 Formula Weight: 255.52 Flash Point: 22°(72°F) Physical Form: Liquid Density: 0.89 Appearance: Yellow Odour: Alcohol-like UN No.: 3274 MDL No.: MFCD00050486


In Sol-Gel synthesis and superconductors

Formula: C₆H₁₄BaO₂
Densidad: 0,89
Pto. de inflamación: 22 °C (72 °F)
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00050486
Núm. CAS: 24363-37-9
UN: 3274
ADR: 3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR