Bismuth tungsten oxide ≥99.9% (metals basis)

Proveedor: Thermo Fisher Scientific

41854.14 41854.22
41854.14EA 160 EUR
41854.14 41854.22
Bismuth tungsten oxide ≥99.9% (metals basis)
Bismuth tungsten oxide

Purity: 99.9% CAS No.: 13595-87-4 Molecular Formula: Bi2(WO4)3 Formula Weight: 1161.48 Physical Form: -200 Mesh Powder Odour: Odourless MDL No.: MFCD00046190

Formula: Bi₂(WO₄)₃
MW: 1161.48 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00046190
Núm. CAS: 13595-87-4
EINECS: 237-033-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR