Sulfur in crude oil standard solutions, Specpure®

Proveedor: Thermo Fisher Scientific

Sulfur in crude oil standard solutions, Specpure®
Normativas Estándares de la industria petrolera

Physical Form: Liquid
Appearance: Black
Odour: Unpleasant
UN No.: UN1267
MDL No.: MFCD00197838

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR