Stainless steel (Fe:Cr:Ni; 70:19:11 wt%), gauze woven from wire, twilled weave 400 mesh woven from 0.028mm (0.0011in) Type 304
Proveedor: Thermo Fisher Scientific
Peligro
42012.CFEA 292 EUR
42012.CF 42012.VA
Stainless steel (Fe:Cr:Ni; 70:19:11 wt%), gauze woven from wire, twilled weave 400 mesh woven from 0.028mm (0.0011in) Type 304
Stainless steel
Molecular Formula: Fe:Cr:Ni; 70:19:11 wt% Physical Form: Wire Cloth Odour: Odourless MDL No.: MFCD00148499