Melting point standards, sodium acetate anhydrous, traceable to primary standards, Supelco®
Proveedor: Merck
42123-5G
42123-5GEA 51.3 EUR
42123-5G
Melting point standards, sodium acetate anhydrous, traceable to primary standards, Supelco®
Normativas Estándar para punto de fusión
Mettler-Toledo Calibration substance ME 30130599, Sodium acetate anhydrous, tracable to primary standards (LGC).
Suitable for dropping point calibration.
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c806a6031591d91f271bbddc28d8bcd89534fcd2&vid=c806a6031591d91f271bbddc28d8bcd89534fcd2&prid=web&clid=SAPDAM