Anti-Lamp1 (LY1C6) Mouse Monoclonal Antibody

Proveedor: Merck Millipore (Oncogene)
428017-100
428017-100EA 692 EUR
428017-100
Anti-Lamp1 (LY1C6) Mouse Monoclonal Antibody
Anticuerpos
Type: Primary
Antigen: Lamp1 (LY1C6)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR