Triiron phosphide ≥99.5% (metals basis)

Proveedor: Thermo Fisher Scientific

43486.09 43486.18
43486.09EA 179 EUR
43486.09 43486.18
Triiron phosphide ≥99.5% (metals basis)
Triiron phosphide

Purity: 99.5% CAS No.: 12023-53-9 Molecular Formula: Fe3P Formula Weight: 198.51 Physical Form: -40 Mesh Powder

Formula: Fe₃P
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00799762
Núm. CAS: 12023-53-9
EINECS: 234-682-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR