VWR® Phosphate Buffered Saline (PBS), Low Phosphate, 10X
Proveedor: VWR Chemicals
437117KEA 271 EUR
437117K
VWR® Phosphate Buffered Saline (PBS), Low Phosphate, 10X
Tampones Tampones para electroforesis Tampones de lavado
Irradiated concentrate of PBS, suitable for use in washing cells after dilution.
  • KCl 2,0 g/L
  • NaCl 70,1 g/L
  • Na₂HPO₄, 2H₂0 12,8 g/L
  • NaH₂PO₄, 1H₂O 4,4 g/L
Añadir 1 ml de PBS X10 a 9 ml de agua para obtener solcuión PBS X1
pH (20 °C): 6,90 - 7,30