Platino nominally 50% on carbon black

Proveedor: Thermo Scientific

Sinónimos: Platinium

43989.14
43989.14EA 0 EUR
43989.14
Platino nominally 50% on carbon black
Platino
Formula: Pt
MW: 195,08 g/mol
Número MDL: MFCD00011179
Núm. CAS: 7440-06-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR