Electronic overhead stirrers
Proveedor: VELP SCIENTIFIC
F201A0150 F201A0152 F201A0151 F201A0155 F201A0156 F201A0157 SA201A0155 SA201A0151
441-0345EA 1070 EUR
441-0345 441-0347 441-0346 441-0348 441-0349 441-0350 441-0413 441-0474
Electronic overhead stirrers
Imánes de agitación Agitadores de varillas
Range of overhead stirrers with electronic adjustment that are equipped with advanced safety systems.
  • Compact, easy to use ergonomic design, units are lightweight, reliable and quiet
  • Progressive start-up prevents liquid from coming out of the vessel
  • Microprocessor controlled motor means constant speed, even when the viscosity of the liquid changes (except ES models)
  • Model BS, PW, DLS, LH and DLH have safety chuck without key, which helps ensure safe use
  • DLS and DLH models have two digital displays showing both actual and set speeds simultaneously