terc-Butanolato de litio ≥99.9% (metals basis)

Proveedor: Thermo Fisher Scientific
Peligro

44133.04 44133.09 44133.18
44133.04EA 60.8 EUR
44133.04 44133.09 44133.18
terc-Butanolato de litio ≥99.9% (metals basis)
terc-Butanolato de litio

Purity: 99.9% CAS No.: 1907-33-1 Molecular Formula: C4H9LiO Formula Weight: 80.06 Physical Form: Powder Sensitivity: Air and Moisture Sensitive Appearance: White to cream Odour: Odourless Melting Point: ca 150° dec. Beilstein Reference: 3620018 UN No.: 3206 MDL No.: MFCD00050479

Formula: C₄H₉LiO
Pto. de fusión: ∼ 150 °C (desc.)
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00050479
Núm. CAS: 1907-33-1
EINECS: 217-611-5
UN: 3206
ADR: 4.2,II

Order Now

ESPECIFICACIONES

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR