Magnetic stirring bar arrester, SP Bel-Art
Proveedor: Bel-Art Products, a Part of SP
F377860000
442-0315EA 50.3 EUR
442-0315
Magnetic stirring bar arrester, SP Bel-Art
Imánes de agitación Herramientas de agitación
Polyethylene (9,5 mm) with radial magnet (Ø 51 mm). Position the arrester against the vessel bottom.
  • Prevents the stirring bar from being poured out with the liquid from a beaker or flask