VWR®, Agitadores de placa magnética, serie Professional
Proveedor: VWR Collection
442-1266EA 1670 EUR
442-1266 442-1271 442-1272 442-1267
VWR®, Agitadores de placa magnética, serie Professional
Imánes de agitación Agitadores magnéticos con placas calefactoras
Designed for applications requiring exceptional accuracy, stability, and repeatability these magnetic hotplate stirrers are equipped with superior heating and mixing capabilities. With temperature ranges up to 500 °C and stirring speeds reaching 1600 rpm, the VWR® magnetic hotplate stirrers are equipped to handle any research, academic and industrial application. Available in two sizes.
  • Excellent temperature uniformity and powerful stirring
  • Easy to read, large LCD displays temperatures, speed and timer settings
  • Cool touch, chemical-resistant housing