Magnetic stirrers, battery powered, SP Bel-Art
Proveedor: Bel-Art Products, a Part of SP
H370170000
442-9460EA 614 EUR
442-9460
Magnetic stirrers, battery powered, SP Bel-Art
Imánes de agitación Agitadores magnéticos
This portable magnetic stirrer is made of PP.
  • Durable and chemical resistant
  • Two D batteries provide 16 days of continuous operation
Ideal for use inside incubators and when electricity is not available.
Información de suministro: Supplied with Spinbar® Octagon Stirbar (BELAF371100138).