Nickel(II) perchlorate hydrate ≥99.998% (metals basis)

Proveedor: Thermo Fisher Scientific
Peligro

44316.09 44316.18
44316.09EA 78 EUR
44316.09 44316.18
Nickel(II) perchlorate hydrate ≥99.998% (metals basis)
Nickel(II) perchlorate hydrate

Purity: 99.998% CAS No.: 13637-71-3 Molecular Formula: Cl2NiO8.xH2O Formula Weight: 257.59(anhy) Physical Form: Crystalline Refractive Index: 1.55 Sensitivity: Hygroscopic Appearance: Green Odour: Odourless Melting Point: 140° UN No.: 1481 MDL No.: MFCD00149804

Formula: Cl₂NiO₈·xH₂O
Pto. de fusión: 140 °C
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00149804
Núm. CAS: 13637-71-3
EINECS: 237-124-1
UN: 1481
ADR: 5.1,II

Order Now

ESPECIFICACIONES

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR