Hot plates, Corning®
Proveedor: Corning
6796-400D 6796-600D 6798-400D 6798-600D
444-1006EA 493.34 EUR
444-1006 444-1009 444-1012 444-1015
Hot plates, Corning®
Calentadores Placas calefactoras
Corning® hotplates with Pyroceram® glass-ceramic top, rugged housing and heating elements and digital temperature display.
  • Microprocessor maintains consistent and repeatable temperature settings from 5 °C (if ambient temperature is 0 °C or lower) up to 550 °C
  • Digital LED temperature display is adjustable in 5 °C increments and blinks until set temperature is reached
  • Optional external temperature controller (444-1004) truly controls temperature inside the vessel
  • Bright LED 'hot' icon lights up when top plate is over 60 °C, even when heat control is turned