Thermoshaker with heating and cooling, Matrix Delta Plus
Proveedor: IKA
30000731 30000717
444-1323EA 2900 EUR
444-1323 444-1322
Thermoshaker with heating and cooling, Matrix Delta Plus
Agitadores de laboratorio Incubadoras de Agitación
The IKA Matrix Delta Plus has heating and cooling functionality and numerous user-friendly features.
  • Large, clear display with a convenient menu structure allows for intuitive operation
  • Programmable ramps allow for automatic execution of heating and heating tasks
  • Secure, quick lock for fast and easy change of inserts and attachments as well as automatic attachment detection
  • Aluminium die-cast housing ensures secure positioning, excellent stability and everyday robustness
  • RS232 and USB interfaces
Matrix Delta Plus units heat and cool to ‒30 °C belowMore Product Information