Supelco® Organic Reference Standard, Bacopaside Ii

Proveedor: Merck

44698-5MG
44698-5MGEA 475 EUR
44698-5MG
Supelco® Organic Reference Standard, Bacopaside Ii
Normativas Estándares orgánicos

Envase: Bottomless glass bottle contents are inside inserted with fused cone.

https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=04a3663d8ff0299c60c9b0d7d4e7ed6e3fe81b92&vid=04a3663d8ff0299c60c9b0d7d4e7ed6e3fe81b92&prid=web&clid=SAPDAM

Order Now

ESPECIFICACIONES

Synonym(s) Pseudojujubogenin 3-O-[α-ʟ-arabinofuranosyl-(1→2)-[β-ᴅ-glucopyranosyl-(1→3)]-β-ᴅ-glucopyranoside]
Environmentally Preferable
M-Clarity quality level MQ100
CAS No. 382146-66-9
Formula C₄₇H₇₆O₁₈
Molecular weight 929,10
Punto de inflamación Not applicable
Condiciones de envío STD
Condiciones de almacenamiento 2...30 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR