Copper, alloy 101, placa, Oxygen-Free High Conductivity (OFHC), Grosor: 2.4 mm (0.093 in)

Proveedor: Thermo Fisher Scientific

45210.HB
45210.HBEA 552 EUR
45210.HB
Copper, alloy 101, placa, Oxygen-Free High Conductivity (OFHC), Grosor: 2.4 mm (0.093 in)
Copper, alloy 101

MDL No.: MFCD00010965

Densidad: 8.94 g/cm³ (20 °C)
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00010965
EINECS: 231-159-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR