Palladium nitrate and magnesium nitrate, matrix modifier solution, Specpure®

Proveedor: Thermo Fisher Scientific

45291.AK
45291.AKEA 205 EUR
45291.AK
Palladium nitrate and magnesium nitrate, matrix modifier solution, Specpure®
Normativas Estándares catiónicos Estándares de multielementos

Pre-mixed matrix modifier are designed for use with graphite furnace atomic absorption (GFAA).


UN No.: UN3264
MDL No.: MFCD09910543

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR