Sulphur in diesel fuel, standard solutions, Specpure®

Proveedor: Thermo Fisher Scientific

Sulphur in diesel fuel, standard solutions, Specpure®
Normativas Estándares de la industria petrolera

Flash Point: 69° (156 °F)
Physical Form: Liquid
Density: 0,6 to 0,9
Appearance: Light brown
Boiling Point: 171 to 315°
MDL No.: MFCD01095233

Order Now

ESPECIFICACIONES

Environmentally Preferable
MDL No. MFCD01095233 MFCD01095233 MFCD01095233
Punto de ebullición 171-315°
Physical form Liquid Liquid Liquid
Apariencia Colourless Light brown
Densidad 0.6-0.9 g/mL
Odour/scent Petroleum-like
Punto de inflamación 69°C(156°F)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR