High-temperature furnaces with lift up door and SiC rod, heating up to 1600 °C, LHTCT models
Proveedor: NABERTHERM
LC014H6ON1 LC014H6ON1G LC014H6ON1GT LC014H6ON1T LC034H6ON5 LC034H6ON5G LC034H6ON5GT LC034H6ON5T LC084H6ON8 LC084H6ON8G LC084H6ON8GT LC084H6ON8T
455-1854EA 7210 EUR
455-1854 455-1855 455-1856 455-1857 455-1862 455-1863 455-1864 455-1865 455-1882 455-1883 455-1884 455-1885
High-temperature furnaces with lift up door and SiC rod, heating up to 1600 °C, LHTCT models
Hornos Hornos
The durability of the SiC rods in periodic use, in combination with their high heating speed, make these high-temperature furnaces good all-rounders in the laboratory. Heating times of 40 minutes to 1400 °C can be achieved, depending on the furnace model and the conditions of use.
  • SiC rod heating
  • Dual shell housing made of textured stainless steel sheets with additional fan cooling for low surface temperature
  • Only fibre materials which are not classified as carcinogenic according to TRGS 905, class 1 or 2 are used
  • Heating rods are easily replaced
  • Lift up