Flask, Digesdahl®
Proveedor: Hach
2312542
458-0048EA 152 EUR
458-0048
Flask, Digesdahl®
Matraces Matraces de fondo plano
Specially designed glass flask for use with Digesdahl® digestion apparatus. Each flask is calibrated individually with a 100 ml (to contain) mark.