Mezclador térmico, ThermoMixer® C
Proveedor: EPPENDORF
5382000.015 5382000.031
460-0223EA 3201 EUR
460-0223 444-1022
Mezclador térmico, ThermoMixer® C
Agitadores de laboratorio Incubadoras de Agitación
The all-in-one for heating/cooling and mixing samples and reactions. The Eppendorf® ThermoMixer C convinces users with its quick and easy-to-exchange SmartBlocks for all common laboratory vessels. There is almost no limit to the variety of applications waiting for you.
  • Mixing, heating, and cooling in one instrument for high flexibility
  • All common vessels and plate formats from 5 µl to 50 ml by different Eppendorf SmartBlocks for standard applications
  • Ideal for cell thawing when using Eppendorf SmartBlock cryo thaw
  • Anti-spill technology prevents lid wetting