Termostatos de refrigeración, serie Alpha
Proveedor: LAUDA
L000643 L000644 L000645 L000646 L000638 L000639 L000640 L000641 L000642
LAUDL000643EA 4870 EUR
LAUDL000643 LAUDL000644 LAUDL000645 LAUDL000646 461-1249 461-1250 461-1251 LAUDL000641 LAUDL000642
Termostatos de refrigeración, serie Alpha
Termostatos con circulación Sistemas de circulación cerrada Circuladores de enfriamiento
The Alpha cooling thermostats have an easy to use control console featuring three button operation and large, clear LED display, ensuring easy navigation through the menu. The thermostats are suitable for use with non flammable liquids (water, water/glycol mixtures). Units can provide both internal and external thermostating. Temperature stability of ±0,05 K gives precise heating and cooling suitable for a wide range of applications. The automatic cooling system enables cooling throughout the entire temperature range of −25 to +100 °C. All cooling thermostats contain a compressor systemMore Product Information