Refrigerated/heating circulators, DYNEO™ DD series
Proveedor: JULABO GmbH
9021704.D 9021701 9021702 9021703 9021704 9021705 9021707 9021701.A 9021702.A 9021703.A 9021708 9021704.A 9021705.A 9021707.A 9021708.A 9021701.D 9021702.D 9021703.D 9021705.D 9021707.D 9021708.D 9021713.N1 9021713.N1.A 9021713.N1.D 9021713.S1 9021713.S1.A 9021713.S1.D 9021714.N1 9021714.N1.A 9021714.N1.D 9021714.S1 9021714.S1.A 9021714.S1.D 9021716.N1 9021716.N1.A 9021716.N1.D 9021715.N1 9021715.N1.A 9021715.N1.D 9021717.N1 9021717.N1.A 9021717.N1.D 9021701.N1 9021701.N1.A 9021701.N1.D 9021728.N1 9021728.N1.A 9021728.N1.D
461-1418EA 6899 EUR
461-1418 461-1388 461-1389 461-1390 461-1391 461-1392 461-1394 461-1401 461-1402 461-1403 461-1395 461-1404 461-1405 461-1407 461-1408 461-1415 461-1416 461-1417 461-1419 461-1421 461-1422 JULA9021713.N1 JULA9021713.N1.A JULA9021713.N1.D JULA9021713.S1 JULA9021713.S1.A JULA9021713.S1.D JULA9021714.N1 JULA9021714.N1.A JULA9021714.N1.D JULA9021714.S1 JULA9021714.S1.A JULA9021714.S1.D JULA9021716.N1 JULA9021716.N1.A JULA9021716.N1.D JULA9021715.N1 JULA9021715.N1.A JULA9021715.N1.D JULA9021717.N1 JULA9021717.N1.A JULA9021717.N1.D JULA9021701.N1 JULA9021701.N1.A JULA9021701.N1.D JULA9021728.N1 JULA9021728.N1.A JULA9021728.N1.D
Refrigerated/heating circulators, DYNEO™ DD series
Termostatos con circulación Sistemas de circulación abierta Circuladores de calentamiento / enfriamiento
DYNEO™ DD refrigerated circulators are powerful cooling units suitable for internal or external applications between –50 and +200 °C. The circulators function precisely and reliably even at elevated ambient temperatures up to +40 °C. The multilingual 8,89 cm colour display and unique rotary knob allow straightforward and intuitive operation. Units have ergonomic handles and a built in drain tap for easy and safe drainage.
  • Optimised cooling coil design saves space in the bath tank
  • Powerful and infinitely adjustable pressure pump
  • Easy to switch between