Boiling baths, analogue, clifton, NE1B Series
Proveedor: NICKEL ELECTRO
NE1B-14 NE1B-28
462-0116EA 1810 EUR
462-0116 462-0117
Boiling baths, analogue, clifton, NE1B Series
Baños Baños de ebullición
Designed for applications that require continuous boiling. Baths have a corrosion resistant stainless steel working environment, with heater and safety cut out concealed underneath, all housed in a durable, chemical resistant, powder coated paint finished outer case.
  • Integral constant level device for connection to water supply combats water evaporation and maintains boiling
  • Include 'low' height removable stainless steel shelf to increase working volume
  • Drain outlet on 28 litre model for convenient bath emptying
  • Boiling can be regulated from vigorous to a