VWR® VWB2, Water Baths
Proveedor: VWR Collection
462-0554EA 803 EUR
462-0554 462-0555 462-0556 462-0557 462-0558 462-0559
VWR® VWB2, Water Baths
Baños Baños de agua
Unstirred water baths with double walled, stainless steel (inner) and coated steel sheet (outer) housing. All baths except the 2 litre models feature a drain tap for convenient emptying and are suitable for use with water or alternatively with heat transfer beads. Baths with a capacity of 5 litres or more have a heat transfer bead mode which is activated through the main menu. A drain stopper (plug) is supplied as on optional accessory to ensure heat transfer beads do not get stuck in the drain outlet. Note: Baths are classified Safety Class 1, allowing unattended operation when used withMore Product Information