Dessiccators, vacuum, space saver, SP Bel-Art
Proveedor: Bel-Art Products, a Part of SP
F420220000 F420100000 F420120000 F420200000 F420250000 F420270000
467-2233EA 226 EUR
467-2233 467-2230 467-2231 467-2232 467-2234 467-0135
Dessiccators, vacuum, space saver, SP Bel-Art
Desecadores Desecadores de plástico
Degas liquids quickly and easily in the laboratory or classroom with this small plastic vacuum desiccator. These strong, shatterproof units provide dry storage and protection for sensitive products.
  • Lighter and does not chip like glass
  • Holds a full vacuum (760 mm Hg) at room temperature for 24 hours
  • White polypropylene bottom models can support incandescent crucibles on Minerit HD high heat ceramic desiccator plates
  • Clear polycarbonate bottom models offer increased viewing visibility from more angles
These dessicators feature a tapered polypropyleneMore Product Information