Accessories for ULT freezer TSX400V
Proveedor: Thermo Scientific
FCR567TSX FCR400TSX FCRP400TSX SK400TSX
HERAFCR567TSXEA 2020 EUR
HERAFCR567TSX 471-1171 471-1172 471-1173
Accessories for ULT freezer TSX400V
Congeladores Ultracongeladores
Record seven days of temperature performance at temperatures to −150 °C using Thermo Scientific™ chart recorders for ultra-low temperature freezers. Recorders use 150 mm chart paper and are available as factory or field installed.
  • Seven-day recorder
  • Field or factory installed
  • Field-installed units must be installed by a qualified professional
Order Now
ESPECIFICACIONES
Environmentally Preferable
Referencia HERAFCR567TSX 471-1171 471-1172 471-1173