Dimetil sulfóxido ≥99.9% ACS, Riedel-de Haën™

Proveedor: Honeywell Chemicals
Atención

Sinónimos: DMSO

472301-4L 472301-4X4L 472301-6X500ML
472301-4LEA 705 EUR
472301-4L 472301-4X4L 472301-6X500ML
Dimetil sulfóxido ≥99.9% ACS, Riedel-de Haën™
Dimetil sulfóxido

ACS GENERAL USE SOLVENTS


  • Value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • Suitable for less demanding applications such as routine isocratic HPLC
  • Variations and decrease issues traceable to solvent contamination
  • All solvents are tested to meet ACS general use specifications

Formula: (CH₃)₂SO
Pto. Ebullición: 189 °C
Pto. de fusión: 18,5 °C
Densidad: 1,104 g/cm³ (20 °C)
Pto. de inflamación: 28,5 %(V)
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00002089
Núm. CAS: 67-68-5
EINECS: 200-664-3
Índice Merck: 13,03285
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3ec1573e80c11ddf9254283ef76abf5fb6539c7e&vid=157e2a24c93041cbbb69833d226195e333b07665&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance complying
Assay (GC) Min. 99.90 %
Water (Karl Fischer) Max. 0.1 %
Non-volatile matter Max. 0.01 %
Acid (by titration) Max. 0.001 meq/g
IR spectrum complying
Refractive Index n20/D 1.479 (lit.)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR