WHEATON® FTA® Nucleic Acid Collection Storage Boxes

Proveedor: DWK Life Sciences
W651611 W651612
479-2126EA 42.8 EUR
479-2126 479-2127
WHEATON® FTA® Nucleic Acid Collection Storage Boxes
Cajas Criocajas
Storage boxes provide an ideal method for batching/storing FTA® Nucleic acid collection pouches.

  • Water repellent fiberboard box material allows for longer durability
  • For use with small or large FTA cards
  • Accommodates two rows of 76,2 mm FTA card pouches or one row of 152,4 FTA card pouches
  • Removable divider for use with smaller pouches
  • For use at RT, +4, −20 °C or colder

Storage boxes are ideal for nucleic acid collection.
Order Now

ESPECIFICACIONES

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR