Nunc™ Internally-Threaded Universal Tubes and Rack System
Proveedor: Thermo Fisher Scientific
374500 374502 374511 374513 374512 374510 374500-BR 374501-BR 374510-BR 374511-BR
479-1196EA 519 EUR
479-1196 479-1198 479-1367 479-1368 479-1380 479-1379 479-2229 479-2230 479-2232 479-2233
Nunc™ Internally-Threaded Universal Tubes and Rack System
Viales
Virgin Class VI medical grade PP. Tubes and rack are ideal for both lower-throughput benchtop use and high-throughput automation use. Permanently bonded, unique 2D barcode laser-etched onto the base of the tube to securely identify and track samples.
  • All barcodes are checked against the database of previously assigned codes to prevent duplicates
  • Every tube is leak tested
  • Externally threaded single-piece caps with moulded-in gaskets that cannot be over-tightened manually or with automated capping tools
  • DNA-, DNase-, RNase- and endotoxin-free
  • Temperature