Bovine mucin (from Submaxillary glands)
Proveedor: Merck Millipore Calbiochem
499643-500
499643-500EA 310 EUR
499643-500
Bovine mucin (from Submaxillary glands)
Proteínas y Péptidos
Native mucin from bovine submaxillary gland is a high molecular weight glycoprotein and its major constituent of saliva, gastrointestinal juices and other secretions.
  • Lyophilised form
  • Hygroscopic
  • Standard handling
Mucin exists as an aggregated complex in excess of 106 molecular weight. It is a rich source of O-linked oligosaccharide chains and it is suitable as a substrate for neuraminidase.