Anti-CD45 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: HI30]

Proveedor: Tonbo Biosciences
50-0459-T025 50-0459-T100
50-0459-T025EA 26.7 EUR
50-0459-T025 50-0459-T100
Anti-CD45 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: HI30]
Anticuerpos
The HI30 antibody reacts with human CD45, one of the most abundant hematopoietic markers and one that is expressed on all leukocytes (the Leukocyte Common Antigen, LCA). CD45 is a protein tyrosine phosphatase existing in several isoforms, each being generated and expressed in cell-specific patterns. With its broad cell distribution, CD45 is critical for many leukocyte functions, regulating signal transduction and cell activation associated with the T cell receptor, B cell receptor, and IL-2 receptor. Other forms of CD45, with restricted cellular expression, include CD45R (B220), CD45RA, CD45More Product Information
Order Now

ESPECIFICACIONES

Simbolo antígeno PTPRC
Environmentally Preferable
Nombre del antígeno Protein tyrosine phosphatase receptor type C
Sinónimos de antígenos CD45R|B220|CD45|GP180|T200|L-CA|LY5|receptor type C|LCA|protein tyrosine phosphatase
Tipo de anticuerpo Primario
Clonalidad Monoclonal
Conjugado PE (Phycoerythrin)
Clon HI30
Reactividad Chimpanzee,Human
Huésped Mouse
ID del gen 5788
Isotipo IgG1 kappa
Citometría de flujo Yes
Temperatura de almacenamiento 2...8 °C, protected from light
Tested applications Flow Cytometry

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR