Anti-LPAM1 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: DATK32]

Proveedor: Tonbo Biosciences
50-5887-U100 50-5887-U025
50-5887-U100EA 141 EUR
50-5887-U100 50-5887-U025
Anti-LPAM1 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: DATK32]
Anticuerpos
The DATK32 antibody is specific for a combinatorial epitope of the mouse integrin alpha 4/beta 7 (LPAM-1) heterodimer. LPAM-1 is expressed on most mature lymphocytes and sub-populations of thymocytes and bone marrow cells. It is involved in lymphocyte adhesion and migration and is a receptor for several extracellular matrix proteins including VCAM-1, fibronectin, and MAdCAM-1.

DATK32 is reported to induce LPAM-1 dependent lymphocyte aggregation and block adhesion to VCAM-1, fibronectin, and MAdCAM-1.

Type: Primary
Antigen: LPAM1 (LPAM-1)
Clonality: monoclonal
Clone: DATK32
Conjugation: PE (More Product Information
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR