VWR®, Test Sieves, 150×50 mm
Proveedor: VWR Collection
510-0578EA 182 EUR
510-0578 510-0579 510-0580 510-0581 510-0582 510-0583 510-0584 510-0585 510-0586 510-0587 510-0588 510-0589 510-0590 510-0591 510-0592 510-0593 510-0594 510-0595 510-0596 510-0597 510-0598 510-0599 510-0600 510-0601 510-0602 510-0603 510-0604 510-0605 510-0606 510-0607 510-0554 510-0555 510-0556 510-0557 510-0558 510-0559 510-0560 510-0561 510-0562 510-0563 510-0564 510-0565 510-0566 510-0567 510-0568 510-0569 510-0570 510-0571 510-0572 510-0573 510-0574 510-0575 510-0576 510-0577
VWR®, Test Sieves, 150×50 mm
Tamices Tamices de ensayo
Frame: Stainless steel; woven wire cloth: Stainless steel, ISO 3310, ASTM E11, BS 410, AFNOR.
Optional: Inspection certificate of calibration 3.1 according to DIN EN 10204 with measuring results of a specific test.
Sieves and accessories of other dimensions are available on request.
Please go to website for a full overview.
All sieves are supplied with serial number and a certificate of compliance with the order 2.1 according to DIN EN 10204. Further certificates and calibration services are available. Contact your local Avantor sales office for further details.
Información deMore Product Information