VWR®, Tamices de prueba, 50 × 25 mm
Proveedor: VWR Collection
510-1792EA 128 EUR
510-1792 510-1828 510-1829 510-1793 510-1794 510-1795 510-1796 510-1797 510-1798 510-1799 510-1800 510-1801 510-1802 510-1803 510-1804 510-1805 510-1806 510-1807 510-1808 510-1809 510-1810 510-1811 510-1812 510-1813 510-1814 510-1815 510-1816 510-1817 510-1818 510-1819 510-1820 510-1821 510-1822 510-1823 510-1824 510-1825 510-1826 510-1827 510-1830 510-1831 510-1832 510-1833 510-1834 510-1835 510-1836 510-1837 510-1838 510-1839 510-1840 510-1841 510-1842 510-1843 510-1844 510-1845 510-1846 510-1847
VWR®, Tamices de prueba, 50 × 25 mm
Tamices Tamices de ensayo
Frame: Stainless steel; woven wire cloth: Stainless steel, ISO 3310, ASTM E11, BS 410, AFNOR.
Optional: Inspection certificate of calibration 3,1 according to DIN EN 10204 with measuring results of a specific test.
Sieves and accessories of other dimensions are available on request. Please go to website for a full overview.
All sieves are supplied with serial number and a certificate of compliance with the order 2,1 according to DIN EN 10204. Further certificates and calibration services are available. Contact your local Avantor sales office for further details.
Información deMore Product Information