VWR®, Test Sieves, 350×60 mm, with Square Holes
Proveedor: VWR Collection
510-3430EA 313 EUR
510-3430 510-3431 510-3432 510-3433 510-3434 510-3435 510-3436 510-3437 510-3438 510-3439 510-3440 510-3441 510-3442 510-3443 510-3444 510-3445 510-3446 510-3447 510-3448 510-3449 510-3450 510-3451 510-3452 510-3453 510-3454 510-3455 510-3456 510-3457 510-3458 510-3459 510-3460 510-3461 510-3462 510-3463 510-3464 510-3465 510-3466 510-3467 510-3468 510-3469 510-3470
VWR®, Test Sieves, 350×60 mm, with Square Holes
Tamices Tamices de ensayo
Frame: Stainless steel; perforated plates: Galvanised steel, ISO 3310-2, ASTM E323, BS 410, AFNOR.
Optional: Inspection certificate of calibration 3.1 according to DIN EN 10204 with measuring results of a specific test.
Sieves and accessories of other dimensions are available on request.
Please go to website for a full overview.
All sieves are supplied with serial number and a certificate of compliance with the order 2.1 according to DIN EN 10204. Further certificates and calibration services are available. Contact your local Avantor sales office for further details.
InformaciónMore Product Information