VWR®, Test Sieves, 250×55 mm
Proveedor: VWR Collection
510-3739EA 185 EUR
510-3739 510-3740 510-3741 510-3742 510-3743 510-3744 510-3745 510-3746 510-3747 510-3748 510-3749 510-3750 510-3751 510-3752 510-3753 510-3754 510-3755 510-3756 510-3757 510-3758 510-3759 510-3760 510-3761 510-3762 510-3763 510-3764 510-3765 510-3766 510-3767 510-3768 510-3769 510-3770 510-3771 510-3772 510-3773 510-3774 510-3775 510-3776 510-3777 510-3778 510-3779 510-3780 510-3781 510-3782 510-3783 510-3784 510-3785 510-3786 510-3787 510-3788 510-3789 510-3790 510-3791 510-3792 510-3793 510-3794 510-3795 510-3796 510-3797 510-3798 510-3799 510-3800 510-3801 510-3802 510-3803 510-3804 510-3805 510-3806 510-3807 510-3808 510-3809 510-3810 510-3811 510-3812 510-3813 510-3715 510-3716 510-3717 510-3718 510-3719 510-3720 510-3721 510-3722 510-3723 510-3724 510-3725 510-3726 510-3727 510-3728 510-3729 510-3730 510-3731 510-3732 510-3733 510-3734 510-3735 510-3736 510-3737 510-3738
VWR®, Test Sieves, 250×55 mm
Tamices Tamices de ensayo
Frame: Stainless steel; woven wire cloth: Stainless steel, ISO 3310, ASTM E11, BS 410, AFNOR.
Optional: Inspection certificate of calibration 3.1 according to DIN EN 10204 with measuring results of a specific test.
Sieves and accessories of other dimensions are available on request.
Please go to website for a full overview.
All sieves are supplied with serial number and a certificate of compliance with the order 2.1 according to DIN EN 10204. Further certificates and calibration services are available. Contact your local Avantor sales office for further details.
Información deMore Product Information