Cassettes, ultrafiltration, Sartocon® Slice
Proveedor: Sartorius
3051467901E-SG 3051463901E-SG 3051465001E-SG 3051466801E-SG 3051443901E-SG 3051462901E-SG 3051463901E-SW
512-0079EA 972 EUR
512-0079 512-0075 512-0076 512-0078 SARP3051443901E-SG SARP3051462901E-SG SARP3051463901E-SW
Cassettes, ultrafiltration, Sartocon® Slice
Filtros Casetes de flujo tangencial
Sartocon® Slice cassettes are made of the same materials and construction and, therefore, the identical flow path length as the Sartocon cassettes, used for larger-scale production. The scale-up and the scale-down is perfectly linear throughout the range of applications, from cell harvesting to protein purification and concentration.
  • Excellent chemical compatibility and wide pH-range
  • Autoclavable or in-line steamable
  • Available with Hydrosart® or PESU membranes
Hydrosart® membrane
Hydrosart® membranes are stabilised cellulose based membranes that haveMore Product Information