Grade 113 qualitative filter papers, Whatman™
Proveedor: Whatman products (Cytiva)
1113-090 1113-110 1113-125 1113-150 1113-185 1113-240 1113-320 1113-500 1113-917 1113-8768
512-0391EA 20.61 EUR
512-0391 512-0481 512-0298 512-0299 512-0300 512-0301 512-0482 512-0483 512-0649 WHAT1113-8768
Grade 113 qualitative filter papers, Whatman™
Filtros Papeles de filtros Papel de filtro cualitativo
Ultra high loading capacity with a particle retention well suited for use with coarse or gelatinous precipitates.
  • Fastest flow rate of the qualitative grades
  • Creped surface
  • Thickest filter paper in the qualitative range
These extremely strong filter papers have a high wet strength due to the addition of a small quantity of chemically stable resin. Normal qualitative applications will not introduce any significant impurities into the filtrate. The resins do, however, contain nitrogen so these grades should not be used in Kjeldahl estimations, etc. SomeMore Product Information